Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004628.4 | NP_004619.3 | 9 | 1923 | ||
NR_027299.1 | 8 | 1799 | ||||
XM_011534093.2 | XP_011532395.1 | 9 | 1880 | |||
GenBank mRNA | AK222844.1 | - | 8 | 1616 | ||
AK289761.1 | - | 9 | 1852 | |||
AK295711.1 | - | 9 | 1741 | |||
AK311039.1 | - | 8 | 1729 | |||
BC016620.1 | - | 9 | 1834 | 4509957 | ||
D21089.1 | - | 9 | 1924 | |||
EU530523.1 | - | 1 | 376 | |||
EU530524.1 | - | 1 | 264 | |||
EU530525.1 | - | 1 | 308 | |||
EU530526.1 | - | 1 | 264 | |||
EU530527.1 | - | 1 | 308 | |||
EU530532.1 | - | 1 | 279 | |||
HM113488.1 | - | 9 | 1823 | |||
KJ534962.1 | - | 9 | 1819 | |||
KJ535085.1 | - | 8 | 1696 | |||
X65024.1 | - | 8 | 1659 |