Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007641.1 | NP_001007642.1 | 1 | 256 | ||
GenBank mRNA | AY461427.1 | - | 1 | 256 | ||
BC168142.1 | - | 284 | 9100645 | |||
FQ210593.1 | - | 295 |