Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPNPEP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003399.5 | NP_003390.4 | 10 | 1089 | ||
GenBank mRNA | BC126174.1 | - | 10 | 1006 | 8991890 | |
BC143901.1 | - | 10 | 1006 | 9052419 | ||
BF830085.1 | - | 4 | 334 | |||
U90724.2 | - | 10 | 1109 |