Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Orc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199092.1 | NP_954523.1 | 4 | 284 | ||
XM_006234150.2 | XP_006234212.1 | 6 | 448 | |||
XM_006234151.2 | XP_006234213.1 | 5 | 374 | |||
XM_006234152.2 | XP_006234214.1 | 7 | 544 | |||
XM_006234154.2 | XP_006234216.1 | 4 | 626 | |||
XM_008761704.1 | XP_008759926.1 | 4 | 626 | |||
XM_008761705.1 | XP_008759927.1 | 6 | 426 | |||
XM_008761706.1 | XP_008759928.1 | 5 | 352 | |||
XM_008761707.1 | XP_008759929.1 | 5 | 744 | |||
XM_008761708.1 | XP_008759930.1 | 5 | 352 | |||
GenBank mRNA | BC060516.1 | - | 4 | 284 | 5598062 | |
FQ213311.1 | - | 408 |