Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003400.3 | NP_003391.1 | 11 | 1641 | ||
XM_005264544.1 | XP_005264601.1 | 10 | 1618 | |||
XM_005264546.1 | XP_005264603.1 | 9 | 749 | |||
XM_006712094.2 | XP_006712157.1 | 12 | 1784 | |||
XM_011533097.1 | XP_011531399.1 | 11 | 950 | |||
XM_011533098.1 | XP_011531400.1 | 11 | 1618 | |||
XM_011533099.2 | XP_011531401.1 | 11 | 2591 | |||
GenBank mRNA | AK289920.1 | - | 11 | 1320 | ||
AK299039.1 | - | 2 | 43 | |||
AK312426.1 | - | 11 | 1079 | |||
AK316481.1 | - | 2 | 57 | |||
AL833550.1 | - | 12 | 2580 | |||
BC032847.2 | - | 12 | 1042 | 5267242 | ||
BX647758.1 | - | 11 | 1623 | |||
CR749840.1 | - | 12 | 1415 | |||
D89729.1 | - | 10 | 931 | |||
Y08614.2 | - | 11 | 951 |