Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr517 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000317.1 | NP_001000317.1 | 1 | 499 |
Gene Symbol: Olr518 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000676.1 | NP_001000676.1 | 1 | 457 |
Set Membership: |
G-protein coupled receptor |