Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000321.1 | NP_001000321.1 | 1 | 520 |
Gene Symbol: LOC100910165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234489.1 | XP_006234551.1 | 520 |
Gene Symbol: Olr546 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001054.1 | NP_001001054.1 | 1 | 520 |
Gene Symbol: Olr550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000322.1 | NP_001000322.1 | 1 | 520 |