Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003404.4 | NP_003395.1 | 5 | 841 | ||
NM_139323.3 | NP_647539.1 | 4 | 746 | |||
XM_017028039.1 | XP_016883528.1 | 5 | 645 | |||
GenBank mRNA | AB209759.1 | - | 3 | 4646 | ||
AB451260.1 | - | 3 | 455 | |||
AB451386.1 | - | 3 | 455 | |||
AF064847.1 | - | 5 | 765 | |||
AK292717.1 | - | 5 | 734 | |||
AL834170.1 | - | 4 | 625 | |||
BC001359.2 | - | 4 | 636 | 3051079 | ||
EU794580.1 | - | 5 | 737 | |||
X57346.1 | - | 5 | 827 |