Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003405.3 | NP_003396.1 | 2 | 606 | ||
GenBank mRNA | AK307750.1 | - | 3 | 730 | ||
AK312650.1 | - | 2 | 450 | |||
AY007132.1 | - | 1 | 193 | |||
BC003047.1 | - | 2 | 541 | 3543571 | ||
CR456612.1 | - | 2 | 409 | |||
L20422.1 | - | 2 | 374 | |||
S80794.1 | - | 2 | 556 | |||
X57345.1 | - | 1 | 289 | |||
X78138.1 | - | 2 | 565 |