Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr713 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000360.1 | NP_001000360.1 | 1 | 238 |
Gene Symbol: Olr711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000625.1 | NP_001000625.1 | 1 | 238 |
Gene Symbol: Olr715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000623.1 | NP_001000623.1 | 1 | 238 |