Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YWHAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003405.3 | NP_003396.1 | 2 | 348 | ||
GenBank mRNA | AK307750.1 | - | 3 | 472 | ||
BC003047.1 | - | 2 | 283 | 3543571 | ||
CR456612.1 | - | 2 | 151 | |||
L20422.1 | - | 2 | 116 | |||
S80794.1 | - | 2 | 298 | |||
X57345.1 | - | 1 | 31 | |||
X78138.1 | - | 2 | 307 |