Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr763 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_230402.2 | XP_230402.2 | 1 | 839 |
Gene Symbol: RGD1566059 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234699.1 | XP_006234761.1 | 2 | 875 |