Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralgapa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762288.1 | XP_008760510.1 | 24 | 3346 | ||
XM_008762289.1 | XP_008760511.1 | 23 | 3205 |