Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plagl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106528.1 | NP_001099998.1 | 3 | 607 | ||
XM_006235285.2 | XP_006235347.1 | 3 | 651 |