Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sun5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106530.1 | NP_001100000.1 | 9 | 801 | ||
XM_008762312.1 | XP_008760534.1 | 11 | 972 |