Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbfa2t2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168542.1 | NP_001162014.1 | 8, 9 | 1350 | ||
XM_006235311.2 | XP_006235373.1 | 8, 9 | 1334 | |||
XM_006235312.2 | XP_006235374.1 | 9, 10 | 1362 | |||
XM_008762313.1 | XP_008760535.1 | 8, 9 | 1438 | |||
GenBank mRNA | FQ222452.1 | - | 8, 9 | 1366 |