Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC103691956 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762450.1 | XP_008760672.1 | 2 | 261 |
Gene Symbol: Fer1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106534.2 | NP_001100004.2 | 21 | 2487 |