Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100885.1 | NP_001094355.1 | 2 | 344 | ||
XM_006235401.1 | XP_006235463.1 | 3 | 460 | |||
XM_006235402.2 | XP_006235464.1 | 3 | 474 | |||
GenBank mRNA | BC158548.1 | - | 3 | 423 | 9036351 | |
FQ215756.1 | - | 3 | 397 | |||
FQ219977.1 | - | 3 | 401 | |||
FQ220245.1 | - | 3 | 401 | |||
FQ220303.1 | - | 3 | 401 | |||
FQ220687.1 | - | 3 | 403 | |||
FQ227515.1 | - | 3 | 404 | |||
FQ229420.1 | - | 3 | 401 | |||
FQ229727.1 | - | 3 | 404 | |||
FQ229762.1 | - | 3 | 403 | |||
FQ229855.1 | - | 3 | 437 | |||
FQ230056.1 | - | 3 | 404 | |||
FQ230407.1 | - | 3 | 417 | |||
FQ231493.1 | - | 3 | 417 | |||
FQ231820.1 | - | 3 | 436 | |||
FQ232091.1 | - | 3 | 397 | |||
S77900.1 | - | 2 | 344 |