Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006235401.1 | XP_006235463.1 | 4 | 1033 | ||
XM_006235402.2 | XP_006235464.1 | 4 | 1047 | |||
GenBank mRNA | BC158548.1 | - | 4 | 996 | 9036351 | |
FQ215756.1 | - | 4 | 970 | |||
FQ220245.1 | - | 4 | 974 | |||
FQ220303.1 | - | 4 | 974 | |||
FQ227515.1 | - | 4 | 977 | |||
FQ229420.1 | - | 4 | 974 | |||
FQ229727.1 | - | 4 | 977 | |||
FQ229762.1 | - | 4 | 976 | |||
FQ229855.1 | - | 4 | 1010 | |||
FQ230056.1 | - | 4 | 977 | |||
FQ230407.1 | - | 4 | 988 | |||
FQ231820.1 | - | 4 | 1009 | |||
FQ232091.1 | - | 4 | 970 |