Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wfdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106541.1 | NP_001100011.1 | 4 | 740 | ||
XM_006235533.2 | XP_006235595.1 | 4 | 545 |