Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
Zmynd8
Gene Name:
zinc finger, MYND-type containing 8
Gene Aliases:
Prkcbp1, spikar
Chromosome Location:
Chr.3: 162480766 - 162594166 on Build Rnor_6.0
UniGene ID:
Rn.219272
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s150109
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Zmynd8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100838.1
NP_001094308.1
9
845
XM_006235536.1
XP_006235598.1
10
878
XM_006235537.1
XP_006235599.1
9
866
XM_006235538.2
XP_006235600.1
10
939
XM_006235539.2
XP_006235601.1
11
1894
XM_006235540.2
XP_006235602.1
9
2844
XM_006235541.2
XP_006235603.1
10
1881
XM_006235542.2
XP_006235604.1
8
2832
XM_006235543.1
XP_006235605.1
10
803
XM_006235544.1
XP_006235606.1
10
878
XM_006235546.1
XP_006235608.1
10
1461
XM_006235547.2
XP_006235609.1
10
1663
XM_006235549.2
XP_006235611.1
10
1449
XM_006235552.2
XP_006235614.1
9
1437
XM_006235553.2
XP_006235615.1
9
1089
XM_006235554.2
XP_006235616.1
11
1819
XM_006235555.2
XP_006235617.1
10
1663
XM_006235556.2
XP_006235618.1
10
1881
XM_006235557.1
XP_006235619.1
10
878
XM_006235559.2
XP_006235621.1
10
1735
XM_006235560.2
XP_006235622.1
8
2832
XM_006235561.2
XP_006235623.1
10
1806
XM_006235562.1
XP_006235624.1
10
803
XM_006235563.1
XP_006235625.1
10
878
XM_006235564.2
XP_006235626.1
10
1645
XM_006235565.2
XP_006235627.1
10
1735
XM_006235566.1
XP_006235628.1
10
803
XM_006235567.2
XP_006235629.1
10
1645
XM_006235568.1
XP_006235630.1
10
878
XM_006235569.2
XP_006235631.1
10
1717
XM_006235571.2
XP_006235633.1
10
1640
XM_006235572.2
XP_006235634.1
4
493
XM_006235573.2
XP_006235635.1
11
1277
XM_008762478.1
XP_008760700.1
10
2168
XM_008762479.1
XP_008760701.1
12
1959
XM_008762480.1
XP_008760702.1
10
1101
XM_008762481.1
XP_008760703.1
10
901
XM_008762482.1
XP_008760704.1
11
1946
XM_008762483.1
XP_008760705.1
9
791
GenBank mRNA
AB074010.1
-
9
845
AB721962.1
-
8
833
BC083796.1
-
8
727
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline