Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZMYND8 SIRNA Assays ›
Gene Symbol:
Zmynd8
Gene Name:
zinc finger, MYND-type containing 8
Gene Aliases:
Prkcbp1, spikar
Chromosome Location:
Chr.3: 162480766 - 162594166 on Build Rnor_6.0
UniGene ID:
Rn.219272
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
s150110
Purification
Standard
HPLC
HPLC-IVR
Size
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
InVivo | 5 nmol
Availability
Made To Order
Catalog #
4390771
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Zmynd8
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001100838.1
NP_001094308.1
3
233
XM_006235536.1
XP_006235598.1
4
266
XM_006235537.1
XP_006235599.1
3
254
XM_006235538.2
XP_006235600.1
4
327
XM_006235539.2
XP_006235601.1
5
1282
XM_006235540.2
XP_006235602.1
3
2232
XM_006235541.2
XP_006235603.1
4
1269
XM_006235542.2
XP_006235604.1
2
2220
XM_006235543.1
XP_006235605.1
4
266
XM_006235544.1
XP_006235606.1
4
266
XM_006235546.1
XP_006235608.1
4
849
XM_006235547.2
XP_006235609.1
4
1051
XM_006235549.2
XP_006235611.1
4
837
XM_006235552.2
XP_006235614.1
3
825
XM_006235553.2
XP_006235615.1
3
477
XM_006235554.2
XP_006235616.1
5
1282
XM_006235555.2
XP_006235617.1
4
1126
XM_006235556.2
XP_006235618.1
4
1269
XM_006235557.1
XP_006235619.1
4
266
XM_006235559.2
XP_006235621.1
4
1123
XM_006235560.2
XP_006235622.1
2
2220
XM_006235561.2
XP_006235623.1
4
1269
XM_006235562.1
XP_006235624.1
4
266
XM_006235563.1
XP_006235625.1
4
266
XM_006235564.2
XP_006235626.1
4
1108
XM_006235565.2
XP_006235627.1
4
1123
XM_006235566.1
XP_006235628.1
4
266
XM_006235567.2
XP_006235629.1
4
1108
XM_006235568.1
XP_006235630.1
4
266
XM_006235569.2
XP_006235631.1
4
1105
XM_006235571.2
XP_006235633.1
4
1103
XM_006235573.2
XP_006235635.1
5
740
XM_008762478.1
XP_008760700.1
4
1556
XM_008762479.1
XP_008760701.1
6
1347
XM_008762480.1
XP_008760702.1
4
489
XM_008762481.1
XP_008760703.1
4
289
XM_008762482.1
XP_008760704.1
5
1334
XM_008762483.1
XP_008760705.1
3
254
GenBank mRNA
AB074010.1
-
3
233
AB721962.1
-
2
221
BC083796.1
-
2
190
Back To Top
More Information
Gene Ontology Categories:
Process(es)
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-vpmkj:80/100.66.78.144:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline