Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013924.1 | NP_001013946.1 | 5 | 611 | ||
XM_006235817.2 | XP_006235879.1 | 5 | 1037 | |||
XM_006235818.2 | XP_006235880.1 | 5 | 657 | |||
XM_006235820.2 | XP_006235882.1 | 5 | 665 | |||
XM_008762485.1 | XP_008760707.1 | 4 | 2171 | |||
XM_008762486.1 | XP_008760708.1 | 5 | 1041 | |||
XM_008762487.1 | XP_008760709.1 | 6 | 758 | |||
XM_008762488.1 | XP_008760710.1 | 6 | 781 | |||
XM_008762489.1 | XP_008760711.1 | 5 | 680 | |||
XM_008762490.1 | XP_008760712.1 | 6 | 737 | |||
XM_008762491.1 | XP_008760713.1 | 5 | 661 | |||
XM_008762492.1 | XP_008760714.1 | 4 | 2171 | |||
XM_008762493.1 | XP_008760715.1 | 5 | 688 | |||
XM_008762494.1 | XP_008760716.1 | 4 | 2171 | |||
XM_008762495.1 | XP_008760717.1 | 4 | 2171 | |||
GenBank mRNA | BC083707.1 | - | 5 | 611 | 7190542 |