Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015394.4 | NP_056209.2 | 5 | 1864 | ||
GenBank mRNA | AK314784.1 | - | 5 | 2125 | ||
AL110188.1 | - | 3 | 1581 | |||
BC024182.2 | - | 5 | 1865 | 4823159 | ||
X52332.1 | - | 5 | 1346 |