Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mrpl41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013426.1 | NP_001013444.1 | 2 | 297 | ||
GenBank mRNA | BC091293.1 | - | 2 | 297 | 7307323 |
Gene Symbol: RGD1560917 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750552.3 | XP_003750600.1 | 1 | 333 |