Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcn9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106560.1 | NP_001100030.1 | 5 | 457 | ||
XM_008761622.1 | XP_008759844.1 | 5 | 477 | |||
GenBank mRNA | DQ537496.1 | - | 5 | 446 |