Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem8c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134517.1 | NP_001127989.1 | 3 | 578 | ||
XM_008761635.1 | XP_008759857.1 | 3 | 580 | |||
XM_008761636.1 | XP_008759858.1 | 3 | 329 | |||
XM_008761637.1 | XP_008759859.1 | 1 | 50 |