Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam73b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106566.1 | NP_001100036.1 | 9 | 1183 | ||
XM_006233781.1 | XP_006233843.1 | 9 | 1232 | |||
XM_006233782.2 | XP_006233844.1 | 9 | 1248 | |||
GenBank mRNA | FQ212119.1 | - | 9 | 1151 | ||
FQ231217.1 | - | 9 | 1187 |