Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF708 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297560.1 | NP_001284489.1 | 4 | 1669 | ||
NM_001297561.1 | NP_001284490.1 | 3 | 1460 | |||
NM_021269.2 | NP_067092.2 | 4 | 1585 | |||
XM_017027201.1 | XP_016882690.1 | 4 | 1487 | |||
XM_017027202.1 | XP_016882691.1 | 4 | 1610 | |||
XM_017027203.1 | XP_016882692.1 | 2 | 1390 | |||
XM_017027204.1 | XP_016882693.1 | 4 | 1402 | |||
GenBank mRNA | AK128184.1 | - | 4 | 1544 | ||
AK131527.1 | - | 4 | 1585 | |||
AK311130.1 | - | 1 | 312 | |||
BX647526.1 | - | 4 | 1655 |