Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfml2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106572.1 | NP_001100042.1 | 7 | 1696 | ||
XM_006234074.2 | XP_006234136.1 | 8 | 1904 |