Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napepld | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199381.1 | NP_955413.1 | 4 | 1091 | ||
XM_006235908.2 | XP_006235970.1 | 5 | 2513 | |||
XM_006235910.2 | XP_006235972.1 | 5 | 1360 | |||
XM_006235911.2 | XP_006235973.1 | 5 | 1314 | |||
XM_008762638.1 | XP_008760860.1 | 5 | 4762 | |||
XM_008762639.1 | XP_008760861.1 | 5 | 2023 | |||
GenBank mRNA | AB112351.1 | - | 4 | 1091 |