Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001203250.1 | NP_001190179.1 | 4 | 927 | ||
NM_021143.3 | NP_066966.2 | 4 | 936 | |||
GenBank mRNA | AK023094.1 | - | 4 | 830 | ||
AL080125.1 | - | 4 | 808 | |||
BC036714.2 | - | 4 | 854 | 4846514 | ||
BC045814.1 | - | 4 | 771 | 4830061 | ||
X52344.1 | - | 1 | 33 |
Gene Symbol: ZNF625-ZNF20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037802.1 | 8 | 1293 | |||
GenBank mRNA | AK096560.1 | - | 8 | 1293 |