Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chchd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106588.1 | NP_001100058.1 | 2 | 294 | ||
GenBank mRNA | FQ216356.1 | - | 2 | 277 |