Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kel | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191611.1 | NP_001178540.1 | 8 | 912 | ||
XM_006236377.2 | XP_006236439.1 | 9 | 1138 | |||
XM_006236378.1 | XP_006236440.1 | 9 | 1138 | |||
XM_006236379.2 | XP_006236441.1 | 7 | 987 | |||
XM_006236380.2 | XP_006236442.1 | 9 | 1138 | |||
XM_006236381.2 | XP_006236443.1 | 7 | 886 | |||
XM_008762879.1 | XP_008761101.1 | 8 | 991 |