Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSA-14-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182670.2 | NP_872611.2 | 1 | 193 | ||
GenBank mRNA | AY149343.1 | - | 1 | 147 | ||
BC083656.1 | - | 1 | 182 | 7135210 | ||
CK603687.1 | - | 1 | 198 |