Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256279.1 | NP_001243208.1 | 5 | 1909 | ||
NM_001256280.1 | NP_001243209.1 | 3 | 1714 | |||
NM_019591.3 | NP_062537.2 | 4 | 1810 | |||
XM_005266182.3 | XP_005266239.1 | 5 | 1897 | |||
XM_005266183.3 | XP_005266240.1 | 6 | 2100 | |||
XM_011534829.1 | XP_011533131.1 | 5 | 2013 | |||
XM_017019920.1 | XP_016875409.1 | 6 | 2533 | |||
XM_017019921.1 | XP_016875410.1 | 6 | 1996 | |||
XM_017019922.1 | XP_016875411.1 | 5 | 2882 | |||
XM_017019923.1 | XP_016875412.1 | 5 | 2813 | |||
GenBank mRNA | AK000762.1 | - | 1 | 1013 | ||
AK122941.1 | - | 3 | 3923 | |||
AK225296.1 | - | 1 | 1013 | |||
AK291243.1 | - | 4 | 1777 | |||
AL833057.1 | - | 1 | 587 | |||
BC008211.1 | - | 1 | 550 | |||
BC046206.2 | - | 4 | 1711 | 6176427 | ||
BT006963.1 | - | 1 | 315 | |||
EU668356.1 | - | 4 | 1688 |