Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctnna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106598.1 | NP_001100068.1 | 12 | 1918 | ||
XM_008763020.1 | XP_008761242.1 | 3 | 955 |