Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: V1ra14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008898.1 | NP_001008898.1 | 1 | 571 | ||
GenBank mRNA | AY510301.1 | - | 1 | 571 |
Gene Symbol: Vom1r103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167553.1 | NP_001161025.1 | 1 | 541 |
Gene Symbol: Vom1r105 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173113.1 | NP_775136.1 | 2 | 765 | ||
GenBank mRNA | U36785.1 | - | 2 | 765 |
Gene Symbol: Vom1r99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166760.1 | NP_001160232.1 | 1 | 538 |