Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005368.2 | NP_001005368.1 | 3 | 1043 | ||
NM_001324164.1 | NP_001311093.1 | 3 | 1011 | |||
NM_001324165.1 | NP_001311094.1 | 3 | 1149 | |||
NM_001324166.1 | NP_001311095.1 | 3 | 913 | |||
NM_001324167.1 | NP_001311096.1 | 3 | 909 | |||
NM_001324168.1 | NP_001311097.1 | 3 | 1047 | |||
NM_006973.2 | NP_008904.1 | 3 | 905 | |||
XM_017016616.1 | XP_016872105.1 | 2 | 3976 | |||
GenBank mRNA | BC022842.1 | - | 2 | 840 | 5014207 | |
BM127643.1 | - | 1 | 446 | |||
CB067143.1 | - | 1 | 110 | |||
U69645.1 | - | 3 | 881 |
Gene Symbol: ZSCAN23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012455.1 | NP_001012458.1 | 4 | 952 | ||
XM_005248950.2 | XP_005249007.1 | 4 | 950 | |||
XM_011514406.2 | XP_011512708.1 | 4 | 950 | |||
XM_011514408.2 | XP_011512710.1 | 5 | 1137 | |||
XM_011514410.2 | XP_011512712.1 | 4 | 950 | |||
XM_017010479.1 | XP_016865968.1 | 4 | 4166 | |||
GenBank mRNA | AK092117.1 | - | 3 | 782 | ||
AK309235.1 | - | 2 | 269 | |||
AL833347.1 | - | 4 | 917 | |||
BC101557.1 | - | 3 | 854 | 8069063 | ||
BC101559.1 | - | 3 | 854 | 8069065 |