Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034131.1 | NP_001029303.1 | 12 | 1590 | ||
XM_006236954.2 | XP_006237016.1 | 11 | 1316 | |||
XM_006236955.2 | XP_006237017.1 | 11 | 1831 | |||
XM_006236956.1 | XP_006237018.1 | 11 | 1253 | |||
XM_008763148.1 | XP_008761370.1 | 14 | 2332 | |||
XM_008763149.1 | XP_008761371.1 | 14 | 1734 | |||
XM_008763150.1 | XP_008761372.1 | 15 | 1809 | |||
XM_008763151.1 | XP_008761373.1 | 14 | 1926 | |||
XM_008763152.1 | XP_008761374.1 | 13 | 1912 | |||
XM_008763153.1 | XP_008761375.1 | 13 | 2458 | |||
XM_008763154.1 | XP_008761376.1 | 14 | 2329 | |||
XM_008763155.1 | XP_008761377.1 | 14 | 2330 | |||
XM_008763156.1 | XP_008761378.1 | 14 | 2330 | |||
XM_008763157.1 | XP_008761379.1 | 14 | 1728 | |||
XM_008763158.1 | XP_008761380.1 | 14 | 1734 | |||
GenBank mRNA | BC100267.1 | - | 12 | 1590 | 7383433 |