Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam21c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199207.1 | NP_954677.1 | 16 | 1509 | ||
XM_006237147.2 | XP_006237209.1 | 16 | 1533 | |||
XM_006237149.2 | XP_006237211.1 | 16 | 1533 | |||
GenBank mRNA | AY257251.1 | - | 16 | 1509 |