Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam21c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199207.1 | NP_954677.1 | 30, 31 | 3931 | ||
XM_006237147.2 | XP_006237209.1 | 29, 30 | 3892 | |||
XM_006237149.2 | XP_006237211.1 | 29, 30 | 3853 | |||
GenBank mRNA | AY257251.1 | - | 30, 31 | 3931 |