Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klre1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181372.2 | NP_852037.1 | 3 | 322 | ||
XM_006237081.2 | XP_006237143.1 | 5 | 627 | |||
XM_006237082.2 | XP_006237144.1 | 4 | 709 | |||
XM_006237083.2 | XP_006237145.1 | 3 | 352 | |||
XM_008763311.1 | XP_008761533.1 | 4 | 706 | |||
GenBank mRNA | AF486185.1 | - | 3 | 263 | ||
AF486186.1 | - | 3 | 322 | |||
AF486187.1 | - | 4 | 410 | |||
FQ221012.1 | - | 4 | 420 |