Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZSCAN21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145914.2 | NP_666019.1 | 4 | 929 | ||
XM_005250568.4 | XP_005250625.1 | 5 | 2071 | |||
XM_006716113.3 | XP_006716176.1 | 4 | 1910 | |||
XM_017012584.1 | XP_016868073.1 | 5 | 2035 | |||
GenBank mRNA | AF155100.1 | - | 3 | 604 | ||
AK292267.1 | - | 4 | 949 | |||
AK301871.1 | - | 6 | 980 | |||
AK313884.1 | - | 4 | 947 | |||
AL136865.1 | - | 3 | 882 | |||
BC033129.1 | - | 1 | 54 | |||
BC047309.1 | - | 4 | 922 | 5111890 | ||
CR749301.1 | - | 5 | 813 |