Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rrs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079699.1 | NP_001073167.1 | 1 | 619 | ||
GenBank mRNA | BC128746.1 | - | 1 | 619 | 7098944 |