Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufaf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276441.1 | NP_001263370.1 | 7 | 746 | ||
GenBank mRNA | FQ215404.1 | - | 7 | 746 |