Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013089.2 | NP_001013107.1 | 11 | 1167 | ||
XM_006238033.2 | XP_006238095.1 | 11 | 1091 | |||
XM_008763615.1 | XP_008761837.1 | 9 | 1055 | |||
GenBank mRNA | BC098756.1 | - | 11 | 1167 | 7370278 |