Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actl7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011973.1 | NP_001011973.1 | 1 | 452 | ||
XM_008763707.1 | XP_008761929.1 | 2 | 515 | |||
GenBank mRNA | BC082101.1 | - | 1 | 452 | 7134568 |