Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem245 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749945.3 | XP_003749993.1 | 11 | 1867 | ||
XM_006238188.2 | XP_006238250.1 | 11 | 1867 | |||
XM_008763737.1 | XP_008761959.1 | 9 | 1738 | |||
XM_216388.8 | XP_216388.4 | 10 | 1843 |