Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000401.1 | NP_001000401.1 | 1 | 70 |
Gene Symbol: LOC100912327 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238173.2 | XP_006238235.1 | 1 | 154 |
Gene Symbol: LOC103690011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763720.1 | XP_008761942.1 | 1 | 154 |
Gene Symbol: Olr847 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000403.1 | NP_001000403.1 | 1 | 70 |